(±)-2-Methyl-2,4-pentanediol 99+%

Supplier: Thermo Scientific Chemicals
Warning

21945-0500
TS21945-0500EA 62 USD
TS21945-0500
(±)-2-Methyl-2,4-pentanediol 99+%
(±)-2-Methyl-2,4-pentanediol
Formula: C₆H₁₄O₂
Boiling Pt: 197 °C (760 mmHg)
Melting Pt: –40 °C
Density: 0.922 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00004547
CAS Number: 107-41-5
EINECS: 203-489-0
Merck Index: 13,04730

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4280 (20°C, 589 nm)
GC ≥99 %
Color scale ≤15 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR