Sodium dichromate dihydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

21924-0025 AC21924-0010
TS21924-0025EA 224 USD
TS21924-0025 TS21924-0010
Sodium dichromate dihydrate 99%
Sodium dichromate dihydrate
Formula: Na₂Cr₂O₇·2H₂O
Melting Pt: 357 °C
Storage Temperature: Ambient
MDL Number: MFCD00149166
CAS Number: 7789-12-0
EINECS: 234-190-3
UN: 3087
ADR: 5.1,II
Merck Index: 12,08754

Order Now

Specification Test Results

Appearance Orange Crystals or needles
Titration Iodometric ≥98.5 %
pH 3.5 to 3.9 (5 % solution)
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤1000 ppm
Insoluble matter ≤0.01 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR