Calcium chloride 96%, granules for analysis
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Chloro calcium, Calcium dichloride, CaCl2
AC21917-0025
AC21917-1000
AC21917-5000
TS21917-0025EA
209
USD
TS21917-0025
TS21917-1000
TS21917-5000
Calcium chloride 96%, granules for analysis
Calcium chloride
Formula:
CaCl₂ Boiling Pt: >1600 °C (760 mmHg) Melting Pt: 782 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 |
Specification Test Results
Appearance | White Granules |
Titration Complexometric | ≥95 % |
Fluoride (F) | ≤40 ppm |
Sulfate (SO4) | ≤0.2 % |
Arsenic (As) | ≤1 ppm |
Barium (Ba) | ≤0.4 % |
Iron (Fe) | ≤50 ppm |
Lead (Pb) | ≤5 ppm |
Mercury (Hg) | ≤1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...