Calcium sulfate, Drierite™ 8 mesh for drying, with indicator

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum, Calcium sulfate (anhydrous) anhydrite

21909-0020 21909-0050 21909-5000
TS21909-0020EA 291 USD
TS21909-0020 TS21909-0050 TS21909-5000
Calcium sulfate, Drierite™ 8 mesh for drying, with indicator
Calcium sulphate
Formula: CaO₄S
MW: 136.14 g/mol
Melting Pt: 1350 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
UN: 3077
ADR: 9 III
Merck Index: 13,01711

Order Now

Specification Test Results

Appearance Blue Irregular granules
Water ≤0.3 %
Particle size 8 mesh
Composition ≤2 % (CoCl2) Typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR