Calcium sulfate, Drierite™ 4 mesh, without indicator
Supplier: Thermo Scientific Chemicals
Synonyms:
Calcium sulphate (anhydrous) anhydrite, Gypsum, Calcium sulfate (anhydrous) anhydrite
21903-0020
21903-0100
219035000
TS21903-0020EA
137
USD
TS21903-0020
TS21903-0100
TS219035000
Calcium sulfate, Drierite™ 4 mesh, without indicator
Calcium sulphate
Formula:
CaO₄S MW: 136.14 g/mol Melting Pt: 1350 Storage Temperature: Ambient |
MDL Number:
MFCD00010912 CAS Number: 7778-18-9 EINECS: 231-900-3 Merck Index: 13,01711 |
Specification Test Results
Appearance | White to grayish-white Irregular granules |
Water | ≤0.3 % |
Particle size | 4 mesh |
Composition | ≥99.5 % (CaSO4) Typical |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...