7-Amino-4-methylcoumarin 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC
21806-0010
21806-0050
21806-2500
TS21806-0010EA
267
USD
TS21806-0010
TS21806-0050
TS21806-2500
7-Amino-4-methylcoumarin 98%
7-Amino-4-methylcoumarin
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Boiling Pt: 352 Melting Pt: 226 Storage Temperature: Ambient |
MDL Number:
MFCD00006868 CAS Number: 26093-31-2 EINECS: 247-454-8 |
Specification Test Results
Appearance | Yellow to beige to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...