7-Amino-4-methylcoumarin 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC

21806-0010 21806-0050 21806-2500
TS21806-0010EA 267 USD
TS21806-0010 TS21806-0050 TS21806-2500
7-Amino-4-methylcoumarin 98%
7-Amino-4-methylcoumarin
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 352
Melting Pt: 226
Storage Temperature: Ambient
MDL Number: MFCD00006868
CAS Number: 26093-31-2
EINECS: 247-454-8

Order Now

Specification Test Results

Appearance Yellow to beige to brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR