(R)-(+)-1-(1-Naphthyl)ethylamine 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine
21780-0010
21780-0050
TS21780-0010EA
50
USD
TS21780-0010
TS21780-0050
(R)-(+)-1-(1-Naphthyl)ethylamine 99+%
(R)-(+)-1-(1-Naphthyl)ethylamine
Formula:
C₁₂H₁₃N Boiling Pt: 153 °C (11 mmHg) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064114 CAS Number: 3886-70-2 EINECS: 223-425-5 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.6215 to 1.6245 (20 °C, 589 nm) |
GC | ≥99.0 % |
Specific optical rotation | +51° to +59° (20 °C, 589 nm) (c=2, ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...