Magnesium nitrate hexahydrate 99+% for analysis
Supplier: Thermo Scientific Chemicals
AC21756-0010
AC21756-5000
TS21756-0010EA
137
USD
TS21756-0010
TS21756-5000
Magnesium nitrate hexahydrate 99+% for analysis
Magnesium nitrate hexahydrate
Formula:
Mg(NO₃)₂·6H₂O Boiling Pt: 330 °C (1013 hPa) Melting Pt: 89 °C Flash Pt: 93.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149779 CAS Number: 13446-18-9 EINECS: 233-826-7 Merck Index: 13,05697 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Titration Complexometric | ≥99.0 % |
pH | 5.0 to 8.2 (5 % in water at 20°C) |
Chloride (Cl) | ≤10 ppm |
Sulfate (SO4) | ≤50 ppm |
Barium (Ba) | ≤20 ppm |
Calcium (Ca) | ≤100 ppm |
Iron (Fe) | ≤5 ppm |
Strontium (Sr) | ≤20 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...