Lead(IV) oxide 97+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC21753-0025 AC21753-0050 AC21753-1000 AC21753-5000
TS21753-0025EA 811 USD
TS21753-0025 TS21753-0050 TS21753-1000 TS21753-5000
Lead(IV) oxide 97+%, Extra Pure
Lead (IV) oxide
Formula: O₂Pb
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011165
CAS Number: 1309-60-0
EINECS: 215-174-5
UN: 1872
ADR: 5.1,III
Merck Index: 12,05424

Order Now

Specification Test Results

Appearance Brown to black Powder
Titration with Na2S2O3 ≥97.0 %
Chloride (Cl) ≤0.02 %
Sulfate (SO4) ≤0.005 %
Copper (Cu) ≤0.01 %
Iron (Fe) ≤0.01 %
Zinc (Zn) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR