Acetonitrile-[D₃] 99.8 atom % D, AcroSeal™ for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyanomethane-D3, Acetonitrile-d3, Ethannitrile-D3, Trideuteroacetonitrile, Methyl-d3 cyanide, Methyl cyanide-D3, Acetonitrile-[²H₃]

21742-1000
TS21742-1000EA 518 USD
TS21742-1000
Acetonitrile-[D₃] 99.8 atom % D, AcroSeal™ for NMR spectroscopy
Acetonitrile-[D3]
Formula: D₃CCN
MW: 44.03 g/mol
Boiling Pt: 79…81 °C (760 mmHg)
Melting Pt: –46 °C
Density: 0.844 g/cm³ (20 °C)
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
UN: 1648
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 % (Coulometric)
D-Enrichment ≥99.75 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR