Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

217310100
TS217310100EA 47 USD
TS217310100
Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
Chloroform-[D1]

Packaged in 1 ml ampoules

Formula: CDCl₃
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: –64 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR