(3-Glycidoxypropyl)trimethoxysilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(3-Glycidyloxypropyl)trimethoxysilane, 3-Glycidoxypropyltrimethoxysilane, (3-(2,3-Epoxypropoxy)propyl)trimethoxysilane
AC21654-0025
AC21654-0050
AC21654-1000
AC21654-5000
TS21654-0025EA
616
USD
TS21654-0025
TS21654-0050
TS21654-1000
TS21654-5000
(3-Glycidoxypropyl)trimethoxysilane 97%
(3-Glycidoxypropyl)trimethoxysilane
Formula:
C₉H₂₀O₅Si MW: 236.34 g/mol Boiling Pt: 299 Melting Pt: 0 Density: 1.07 g/cm³ Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005144 CAS Number: 2530-83-8 EINECS: 219-784-2 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4280 to 1.4300 (20°C, 589 nm) |
GC | ≥96.0 % |
Color scale | ≤50 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...