Aluminium oxide 99.7+%, extra pure

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

AC21570-0010 AC21570-0050 AC21570-2500
TS21570-0010EA 67 USD
TS21570-0010 TS21570-0050 TS21570-2500
Aluminium oxide 99.7+%, extra pure
Aluminium oxide
Formula: Al₂O₃
Boiling Pt: 2977 °C (760 mmHg)
Melting Pt: 2030 °C
Density: 3.97 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now

Specification Test Results

Appearance White Powder
Sodium oxide (Na2O) ≤0.1 %
Silicon dioxide (SiO2) ≤0.04 %
Aluminium oxide (Al2O3) ≥99.70 %
Iron(III)oxide (Fe2O3) ≤0.03 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR