Molecular sieve A5 (0.5 nm, 5 Å), powder < 50 µm

Supplier: Thermo Scientific Chemicals

Synonyms: Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets

214815000
TS214815000EA 75 USD
TS214815000
Molecular sieve A5 (0.5 nm, 5 Å), powder < 50 µm
Molecular sieve A5 (0.5 nm, 5 Å)
MDL Number: MFCD00131613
CAS Number: 69912-79-4

Order Now

Specification Test Results

Appearance White to off-white Powder
pH 10 to 10.8 (10 % suspension in water)
Residual water ≤1.5 % (550°C, 2 h)
Water adsorption ≥23 % (50%rH, 20°C, 24h)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR