Molecular sieve A4 (0.4 nm, 4 Å), powder <50 µm

Supplier: Thermo Scientific Chemicals

214800010 214800025 214800100 214800250 214805000
TS214800010EA 153 USD
TS214800010 TS214800025 TS214800100 TS214800250 TS214805000
Molecular sieve A4 (0.4 nm, 4 Å), powder <50 µm
Molecular sieve A4 (0.4 nm, 4 Å)
Storage Temperature: Ambient
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR