Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

214550100
TS214550100EA 214 USD
TS214550100
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Dichloromethane-[D2]
Formula: CD₂Cl₂
MDL Number: MFCD00000882
CAS Number: 1665-00-5
UN: 1593

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR