Stains-all 97%, pure

Supplier: Thermo Scientific Chemicals
Warning

21351-0010 21351-0050 21351-2500
TS21351-0010EA 97 USD
TS21351-0010 TS21351-0050 TS21351-2500
Stains-all 97%, pure
Stains-all
Formula: C₃₀H₂₇BrN₂S₂
Storage Temperature: Ambient
MDL Number: MFCD00078788
CAS Number: 7423-31-6

Order Now

Specification Test Results

Appearance Dark grey to dark purple Powder
Infrared spectrum Conforms
UV spectrum Complies
TLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR