Stains-all 97%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
21351-0010
21351-0050
21351-2500
TS21351-0010EA
97
USD
TS21351-0010
TS21351-0050
TS21351-2500
Stains-all 97%, pure
Stains-all
Formula:
C₃₀H₂₇BrN₂S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00078788 CAS Number: 7423-31-6 |
Specification Test Results
Appearance | Dark grey to dark purple Powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
TLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...