Nitromethane 99+% for analysis

Supplier: Thermo Scientific Chemicals
Warning

213370025 213371000 213375000
TS213370025EA 284 USD
TS213370025 TS213371000 TS213375000
Nitromethane 99+% for analysis
Nitromethane
Formula: CH₃NO₂
Boiling Pt: 100…102 °C (760 mmHg)
Melting Pt: –29 °C
Density: 1.12 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00007400
CAS Number: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR