Potassium fluoride 99+% for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC21260-2500
TS21260-2500EA 130 USD
TS21260-2500
Potassium fluoride 99+% for analysis
Potassium fluoride
Formula: KF
Boiling Pt: 1505 °C (1013 hPa)
Melting Pt: 860 °C
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 13,07714

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤200 ppm
Potassium fluosilicate ≤0.1 %
Cadmium (Cd) ≤5 ppm
Cobalt (Co) ≤5 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤5 ppm
Nickel (Ni) ≤5 ppm
Zinc (Zn) ≤5 ppm
Free acid (HF) ≤0.01 %
Free alkali ≤0.01 % (K2CO3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR