Potassium bromide 99+% for biochemistry

Supplier: Thermo Scientific Chemicals
Warning

21258-0010 21258-2500
TS21258-0010EA 279 USD
TS21258-0010 TS21258-2500
Potassium bromide 99+% for biochemistry
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
Merck Index: 13,07701

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Titration Argentometric ≥99.0 %
Heavy metals ≤5 ppm
Chloride (Cl) ≤0.2 %
Iodide (I) ≤0.001 %
Bromate (BrO3) ≤0.001 %
Iodate (IO3) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Barium (Ba) ≤0.002 %
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Sodium (Na) ≤0.02 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR