Magnesium acetate tetrahydrate for biochemistry

Supplier: Thermo Scientific Chemicals

21255-0010 21255-2500
TS21255-0010EA 202 USD
TS21255-0010 TS21255-2500
Magnesium acetate tetrahydrate for biochemistry
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
Melting Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Complexometric ≥98.5 %
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Barium (Ba) ≤0.001 %
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤0.0005 %
Manganese (Mn) ≤0.001 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR