Aluminium oxide 99.99% (trace metals basis), extra pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminium(III) oxide, Alumina C, Aluminum oxide
AC21225-0250
AC21225-1000
TS21225-0250EA
101
USD
TS21225-0250
TS21225-1000
Aluminium oxide 99.99% (trace metals basis), extra pure
Aluminium oxide
Formula:
Al₂O₃ Boiling Pt: 2977 °C (760 mmHg) Melting Pt: 2030 °C Density: 3.97 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 13,00355 |
Specification Test Results
Appearance | White Powder |
Total trace metal impurities | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...