Crystal violet, pure indicator

Supplier: Thermo Scientific Chemicals
Danger

21212-0250 21212-1000 21212-5000
TS21212-0250EA 39 USD
TS21212-0250 TS21212-1000 TS21212-5000
Crystal violet, pure indicator
Crystal Violet
Formula: C₂₅H₃₀ClN₃
Melting Pt: 173 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 13,04406

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Sensitivity (as indicator) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR