Neodymium(III) oxide 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide

AC21158-0250 AC21158-1000
TS21158-0250EA 74 USD
TS21158-0250 TS21158-1000
Neodymium(III) oxide 99.9% (trace metals basis)
Neodymium (III) oxide
Formula: Nd₂O₃
Boiling Pt: 3760 °C (1013 hPa)
Melting Pt: 2270 °C
Density: 7.24 g/cm³
MDL Number: MFCD00011134
CAS Number: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,06450

Order Now

Specification Test Results

Appearance Blue Powder
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR