Silver(I) hexafluorophosphate 98% -6 mesh

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Silver hexafluorophosphate

AC21112-0010 AC21112-0100 AC21112-0250
TS21112-0010EA 71 USD
TS21112-0010 TS21112-0100 TS21112-0250
Silver(I) hexafluorophosphate 98% -6 mesh
Silver (I) hexafluorophosphate
Formula: AgF₆P
Storage Temperature: Ambient
MDL Number: MFCD00003415
CAS Number: 26042-63-7
EINECS: 247-428-6
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance White to gray-beige Powder
Silver (Ag) ≥41.6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR