Deuterium chloride 1M in D₂O 99.8 atom % D, AcroSeal™ for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hydrogen chloride-D1

21060-1000
TS21060-1000EA 268 USD
TS21060-1000
Deuterium chloride 1M in D₂O 99.8 atom % D, AcroSeal™ for NMR spectroscopy
Deuterium chloride
Formula: DCl
MW: 37.45
Storage Temperature: Ambient
MDL Number: MFCD00044642
CAS Number: 7698-05-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Appearance Colorless Liquid
D-Enrichment ≥99.75 %
Strength 0.95 to 1.05 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR