Hexane (mixture of isomers), technical
Supplier: Thermo Scientific Chemicals
|
Danger
|
21059-0250
210590050
210590010
210590025
TS21059-0250EA
513
USD
TS21059-0250
TS210590050
TS210590010
TS210590025
Hexane (mixture of isomers), technical
Hexane (mixture of isomers)
Formula:
H₃C(CH₂)₄CH₃ Boiling Pt: 69 °C Melting Pt: –95 °C Density: 0.659 g/cm³ Flash Pt: –22 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179311 CAS Number: 110-54-3 EINECS: 295-570-2 UN: 1208 ADR: 3,II REACH: 01-2119484651-34 |
|
Specification Test Results
Appearance | Clear liquid |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...