Silver iodide 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

AC21042-0050 AC21042-0250 AC21042-1000
TS21042-0050EA 53 USD
TS21042-0050 TS21042-0250 TS21042-1000
Silver iodide 99%, pure
Silver iodide
Formula: AgI
Boiling Pt: 1506 °C (760 mmHg)
Melting Pt: 557 °C
Density: 5.68 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003412
CAS Number: 7783-96-2
EINECS: 232-038-0
UN: 3077
ADR: 9,III
Merck Index: 12,08659

Order Now

Specification Test Results

Appearance Yellow to yellow-green Powder
Assay ≥98.5 % (ex Ag)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR