Silver perchlorate hydrate 99%

Supplier: Thermo Scientific Chemicals
Danger

21021-1000 AC21021-0050 AC21021-0250
TS21021-1000EA 462 USD
TS21021-1000 TS21021-0050 TS21021-0250
Silver perchlorate hydrate 99%
Silver perchlorate hydrate
Formula: AgClO₄·xH₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00149128
CAS Number: 331717-44-3
EINECS: 232-035-4
UN: 1481
ADR: 5.1,II
Merck Index: 12,08667

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Argentometric ≥98.5 % (on dry substance)
Water ≤10 % (K.F.)
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Iron (Fe) ≤20 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR