Potassium chlorate 99%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

AC20886-0010
TS20886-0010EA 146 USD
TS20886-0010
Potassium chlorate 99%, Extra Pure
Potassium chlorate
Formula: KClO₃
Boiling Pt: 400 °C (1013 hPa)
Melting Pt: 356…368 °C
Storage Temperature: Ambient
MDL Number: MFCD00011361
CAS Number: 3811-04-9
EINECS: 223-289-7
UN: 1485
ADR: 5.1,II
Merck Index: 13,07703

Order Now

Specification Test Results

Appearance White Crystalline powder
Assay ≥98.5 % (KClO3)
Chloride (Cl) ≤2000 ppm
Bromate (BrO3) ≤100 ppm
Chromium (Cr) ≤200 ppm
Iron (Fe) ≤20 ppm
Insoluble matter ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR