Vanadium(V) oxide 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide

AC20799-0025 AC20799-1000 AC20799-5000
TS20799-0025EA 408 USD
TS20799-0025 TS20799-1000 TS20799-5000
Vanadium(V) oxide 98+%
Vanadium (V) oxide
Formula: O₅V₂
Boiling Pt: 1750 °C (760 mmHg)
Melting Pt: 690 °C
Density: 3.35 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011457
CAS Number: 1314-62-1
EINECS: 215-239-8
UN: 2862
ADR: 6.1,III
Merck Index: 13,09987

Order Now

Specification Test Results

Appearance Yellow to orange-brown Powder
Assay ≥98.0 %
Particle size ≤500 µm
Chromium (Cr) ≤200 ppm
Iron (Fe) ≤1000 ppm
Silicon (Si) ≤200 ppm
Sodium oxide (Na2O) ≤300 ppm
Potassium oxide (K2O) ≤300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR