Vanadium(V) oxide 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Vanadium pentoxide, di-Vanadium pentaoxide
AC20799-0025
AC20799-1000
AC20799-5000
TS20799-0025EA
408
USD
TS20799-0025
TS20799-1000
TS20799-5000
Vanadium(V) oxide 98+%
Vanadium (V) oxide
Formula:
O₅V₂ Boiling Pt: 1750 °C (760 mmHg) Melting Pt: 690 °C Density: 3.35 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011457 CAS Number: 1314-62-1 EINECS: 215-239-8 UN: 2862 ADR: 6.1,III Merck Index: 13,09987 |
Specification Test Results
Appearance | Yellow to orange-brown Powder |
Assay | ≥98.0 % |
Particle size | ≤500 µm |
Chromium (Cr) | ≤200 ppm |
Iron (Fe) | ≤1000 ppm |
Silicon (Si) | ≤200 ppm |
Sodium oxide (Na2O) | ≤300 ppm |
Potassium oxide (K2O) | ≤300 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...