Sodium tungstate dihydrate 99+% for analysis

Supplier: Thermo Scientific Chemicals
Warning

AC20762-0025 AC20762-0050 AC20762-1000 AC20762-5000
TS20762-0025EA 1430 USD
TS20762-0025 TS20762-0050 TS20762-1000 TS20762-5000
Sodium tungstate dihydrate 99+% for analysis
Sodium wolframate dihydrate
Formula: Na₂WO₄·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2
EINECS: 236-743-4
Merck Index: 12,08847

Order Now

Specification Test Results

Appearance White Powder or flakes
Assay ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤100 ppm
Arsenic (As) ≤5 ppm
Molybdenum (Mo) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Free alkali (NaOH) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR