Ammonium thiocyanate 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

20650-0025 20650-5000 AC20650-0010 AC20650-0100
TS20650-0025EA 314 USD
TS20650-0025 TS20650-5000 TS20650-0010 TS20650-0100
Ammonium thiocyanate 99+%, Extra Pure
Ammonium thiocyanate
Formula: NH₄SCN
Melting Pt: 149 °C
Density: 1.3 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564

Order Now

Specification Test Results

Appearance White Crystals
Titration Argentometric ≥99.0 % (on dried basis)
Water ≤1.0 %
pH 4.0 to 6.0 (5 % in water)
Iron (Fe) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR