Diboron trioxide 98% -40 mesh

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Boron oxide, Boric anhydride

AC20630-0010 AC20630-0050 AC20630-0250
TS20630-0010EA 94 USD
TS20630-0010 TS20630-0050 TS20630-0250
Diboron trioxide 98% -40 mesh
di-Boron trioxide
Formula: B₂O₃
Boiling Pt: 1860 °C (760 mmHg)
Melting Pt: 450 °C
Density: 2.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 13,01327

Order Now

Specification Test Results

Appearance White Powder
Total trace metal impurities ≤0.2 %
Titration with NaOH ≥97.5 %
Impurity ≤0.01 % (Total halides impurities)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR