di-Sodium tetraborate decahydrate 99.5% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate

AC20595-0010 AC20595-0050 AC20595-0100
TS20595-0010EA 62 USD
TS20595-0010 TS20595-0050 TS20595-0100
di-Sodium tetraborate decahydrate 99.5% for analysis
di-Sodium tetraborate decahydrate
Formula: Na₂B₄O₇·10H₂O
Melting Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00149193
CAS Number: 1303-96-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 13,08662

Order Now

Specification Test Results

Appearance White Crystalline powder
pH 9 to 9.5 (5 % at 20°C)
Titration with HClO4 ≥99.5 %
Chloride (Cl) ≤10 ppm
Phosphate (PO4) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR