Potassium sulfate, anhydrous 99+%, granular for analysis

Supplier: Thermo Scientific Chemicals

AC20594-0025 AC20594-5000
TS20594-0025EA 196 USD
TS20594-0025 TS20594-5000
Potassium sulfate, anhydrous 99+%, granular for analysis
Potassium sulphate
Formula: K₂SO₄
Boiling Pt: 1689 °C (760 mmHg)
Melting Pt: 1067 °C
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Appearance White Granular powder
pH 5.5 to 8.5 (5 % at 25°C)
Titration after Ion exchange ≥99.0 %
Chloride (Cl) ≤10 ppm
Calcium (Ca) ≤200 ppm
Iron (Fe) ≤5 ppm
Sodium (Na) ≤200 ppm
Heavy metals (as Pb) ≤5 ppm
Nitrogen compounds (as N) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR