Potassium dihydrogen phosphate 99+%, Extra Pure

Supplier: Thermo Scientific Chemicals

20592-5000 AC20592-0025 AC20592-0100
TS20592-5000EA 111 USD
TS20592-5000 TS20592-0025 TS20592-0100
Potassium dihydrogen phosphate 99+%, Extra Pure
Potassium dihydrogen phosphate
Formula: KH₂PO₄
Melting Pt: 253 °C
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals
Loss on drying ≤1.0 % (105°C)
pH 4.2 to 4.7 (1 % at 20°C)
Titration with NaOH ≥99.0 % (on dried substance)
Arsenic (As) ≤3 ppm
Lead (Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR