Formamide 99.5% for analysis
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Carbamaldehyde
20582-0010
20582-0025
20582-1000
205820100
TS20582-0010EA
322
USD
TS20582-0010
TS20582-0025
TS20582-1000
TS205820100
Formamide 99.5% for analysis
Formamide
Formula:
HCONH₂ Boiling Pt: 210 °C (760 mmHg) Melting Pt: 2…3 °C Density: 1.133 g/cm³ Flash Pt: 175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007941 CAS Number: 75-12-7 EINECS: 200-842-0 Merck Index: 13,04261 |
|
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Water | ≤0.05 % (Coulometric) |
Sulfated ash | ≤0.005 % |
GC | ≥99.5 % |
Color scale | ≤10 APHA |
Copper (Cu) | ≤1 ppm |
Iron (Fe) | ≤1 ppm |
Zinc (Zn) | ≤1 ppm |
Heavy metals (as Pb) | ≤1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...