Formamide 99.5% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Carbamaldehyde

20582-0010 20582-0025 20582-1000 205820100
TS20582-0010EA 322 USD
TS20582-0010 TS20582-0025 TS20582-1000 TS205820100
Formamide 99.5% for analysis
Formamide
Formula: HCONH₂
Boiling Pt: 210 °C (760 mmHg)
Melting Pt: 2…3 °C
Density: 1.133 g/cm³
Flash Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.05 % (Coulometric)
Sulfated ash ≤0.005 %
GC ≥99.5 %
Color scale ≤10 APHA
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Zinc (Zn) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR