Silver(I) oxide 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: di-Silver oxide, Disilver oxide, Silver oxide

AC20580-0100 AC20580-0500 AC20580-2500
TS20580-0100EA 78 USD
TS20580-0100 TS20580-0500 TS20580-2500
Silver(I) oxide 99+%
Silver(I) oxide
Formula: Ag₂O
MW: 231.74 g/mol
Melting Pt: 800…830 °C
Storage Temperature: Ambient
MDL Number: MFCD00003404
CAS Number: 20667-12-3
EINECS: 243-957-1
UN: 1479
ADR: 5.1 I
Merck Index: 12,08665

Order Now

Specification Test Results

Appearance Black Powder
Elemental Analysis ≥99.0 %
Nitrate (NO3) ≤50 ppm
Sulfate (SO4) ≤100 ppm
Aluminium (Al) ≤10 ppm
Iron (Fe) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR