Magnesium silicate, Florisil® 60-100 mesh for column chromatography

Supplier: Thermo Scientific Chemicals

20545-0010
TS20545-0010EA 321 USD
TS20545-0010
Magnesium silicate, Florisil® 60-100 mesh for column chromatography
Magnesium silicate
Formula: MgO₃Si
Storage Temperature: Ambient
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size 6.9 % (140 mesh) typical


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR