Ethyl methanesulfonate 99%

Supplier: Thermo Scientific Chemicals
Danger

20526-1000 20526-0100 20526-0250 20526-5000
TS20526-1000EA 412 USD
TS20526-1000 TS20526-0100 TS20526-0250 TS20526-5000
Ethyl methanesulfonate 99%
Ethyl methanesulphonate
Formula: C₃H₈O₃S
MW: 124.16 g/mol
Boiling Pt: 213
Melting Pt: –96 °C
Density: 0.92 g/cm³
Flash Pt: 6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007559
CAS Number: 62-50-0
EINECS: 209-734-8
UN: 2333
ADR: 3 II
Merck Index: 13,03859

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4030 to 1.4050 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR