Ethyl methanesulfonate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
20526-1000
20526-0100
20526-0250
20526-5000
TS20526-1000EA
412
USD
TS20526-1000
TS20526-0100
TS20526-0250
TS20526-5000
Ethyl methanesulfonate 99%
Ethyl methanesulphonate
Formula:
C₃H₈O₃S MW: 124.16 g/mol Boiling Pt: 213 Melting Pt: –96 °C Density: 0.92 g/cm³ Flash Pt: 6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007559 CAS Number: 62-50-0 EINECS: 209-734-8 UN: 2333 ADR: 3 II Merck Index: 13,03859 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4030 to 1.4050 (20°C, 589 nm) |
GC | ≥98.5 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...