Kojic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone
20522-0050
20522-0250
20522-1000
20522-5000
TS20522-0050EA
105
USD
TS20522-0050
TS20522-0250
TS20522-1000
TS20522-5000
Kojic acid 99%
Kojic acid
Formula:
C₆H₆O₄ Melting Pt: 151…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006580 CAS Number: 501-30-4 EINECS: 207-922-4 Merck Index: 1305334 |
Specification Test Results
Appearance | White to beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 151 to 156 °C |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...