Iminodiacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2'-Azanediyldiacetic acid

20497-0025 20497-0250 20497-5000
TS20497-0025EA 242 USD
TS20497-0025 TS20497-0250 TS20497-5000
Iminodiacetic acid 98+%
Iminodiacetic acid
Formula: C₄H₇NO₄
Melting Pt: 243 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004280
CAS Number: 142-73-4
EINECS: 205-555-4

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR