N,N-Dimethyl-1,4-phenylenediammonium dichloride 99%

Supplier: Thermo Scientific Chemicals
Danger

20435-0250 20435-1000
TS20435-0250EA 92 USD
TS20435-0250 TS20435-1000
N,N-Dimethyl-1,4-phenylenediammonium dichloride 99%
N,N-Dimethyl-1,4-phenylenediammonium dichloride
Formula: (CH₃)₂NC₆H₄NH₂·2HCl
Melting Pt: 222 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012991
CAS Number: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1,II
Merck Index: 13,03280

Order Now

Specification Test Results

Appearance White to yellow to beige to pink to grey Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless to yellow to purple-brown
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR