N,N-Dimethyl-1,3-phenylenediammonium dichloride 99%

Supplier: Thermo Scientific Chemicals
Danger

20434-0050 20434-0250
TS20434-0050EA 65 USD
TS20434-0050 TS20434-0250
N,N-Dimethyl-1,3-phenylenediammonium dichloride 99%
N,N-Dimethyl-1,3-phenylenediammonium dichloride
Formula: C₈H₁₄Cl₂N₂
Melting Pt: 217 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012971
CAS Number: 3575-32-4
EINECS: 222-693-0
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to beige to grey to purple Powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR